Genkwanin, 7-methoxyapigenin ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 5-Hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-chromen-4-one, 4',5-Dihydroxy-7-methoxyflavone

D11550MG
100264-436EA 102 USD
100264-436
Genkwanin, 7-methoxyapigenin ≥99%, powder
Genkwanin, 7-methoxyapigenin

  • Light yellow fluffy powder
  • Store in a cool, dry place

Formula: C₁₆H₁₂O₅
MW: 284.27 g/mol
Melting Pt: 288…290 °C
MDL Number: MFCD00017452
CAS Number: 437-64-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38d2534a8b0265c6c49e5e920f6d1be6bbf253b9&vid=38d2534a8b0265c6c49e5e920f6d1be6bbf253b9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR