7-Methoxyisoflavone ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7-Methoxy-3-phenyl-4H-chromen-4-one

1959610MG
100264-220EA 75.85 USD
100264-220
7-Methoxyisoflavone ≥99%, powder
7-Methoxyisoflavone

  • Off white shining powder
  • Store in a cool, dry place

Formula: C₁₆H₁₂O₃
MW: 252.27 g/mol
Melting Pt: 156.6…158.8 °C
MDL Number: MFCD00181416
CAS Number: 1621-56-3
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2265b8d74256ed0822d73f1067e004209fe90bc6&vid=2265b8d74256ed0822d73f1067e004209fe90bc6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR