3'-Bromo-5'-chloro-2'-hydroxychalcone ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19440100MG
100264-124EA 44.46 USD
100264-124
3'-Bromo-5'-chloro-2'-hydroxychalcone ≥98%, powder
3'-Bromo-5'-chloro-2'-hydroxychalcone

Orange shining powder


Store in a cool, dry place

Formula: C₁₅H₁₀BrClO₂
MW: 337.60 g/mol
Melting Pt: 144…146.2 °C
MDL Number: MFCD03412366
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10faab22cc46aabfd707802c7dae5e97b89afa65&vid=454c42919f593ab018ed0ed95ad76bece3bca716&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR