Cinnamyl alcohol ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol

026025100GM
100263-762EA 32.69 USD
100263-762
Cinnamyl alcohol ≥98%
Cinnamyl alcohol

  • White, fused solid
  • Store in a cool, dry place

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 256
Melting Pt: 32…35 °C
Flash Pt: 126
MDL Number: MFCD00002921
CAS Number: 104-54-1
Merck Index: 13,02325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddc2fba7f8ad3cbdf334f56ed71e60cb0f73a8b2&vid=ddc2fba7f8ad3cbdf334f56ed71e60cb0f73a8b2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR