D-(-)-Glutamic acid ≥98.5%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid

0255895GM
100263-752EA 35.31 USD
100263-752
D-(-)-Glutamic acid ≥98.5%, crystalline powder
D(-)-Glutamic acid

  • White crystalline powder
  • Store in a cool, dry place

Formula: C₅H₉NO₄
Melting Pt: 205 °C (dec.)
MDL Number: MFCD00063112
CAS Number: 6893-26-1
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d641c67c83f68fe7ee2e364a291f5f12fcf498c&vid=9d641c67c83f68fe7ee2e364a291f5f12fcf498c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR