Solatunine ≥98% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

021652S10MG
100263-548EA 405.38 USD
100263-548
Solatunine ≥98% (by HPLC)
Solatunine
Formula: C₄₅H₇₃NO₁₅
Melting Pt: 266 °C
MDL Number: MFCD00077873
CAS Number: 20562-02-1
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9d1dfd8dfe34ea99cb80afc8275e4c7bdc11f70&vid=e9d1dfd8dfe34ea99cb80afc8275e4c7bdc11f70&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR