4',5,7-Trihydroxy-3'-methoxyflavanone ≥99% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

021118S10MG
100263-494EA 372.69 USD
100263-494
4',5,7-Trihydroxy-3'-methoxyflavanone ≥99% (by HPLC)
4',5,7-Trihydroxy-3'-methoxyflavanone
Formula: C₁₆H₁₄O₆
Melting Pt: 225…227 °C
MDL Number: MFCD00017314
CAS Number: 107657-60-3
UN: 12352100
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR