Hyperosid ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Hyperoside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Hyperin, Quercetin-3-galactoside

021027S10MG
100263-480EA 176.54 USD
100263-480
Hyperosid ≥98%, powder
Hyperosid

  • Yellow powder
  • To be kept in a desiccator

Formula: C₂₁H₂₀O₁₂
MW: 464.38 g/mol
Melting Pt: 220…222 °C
MDL Number: MFCD00016933
CAS Number: 482-36-0
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d049c01e3ebb68d64f950433f250e6d9fc3106b8&vid=d049c01e3ebb68d64f950433f250e6d9fc3106b8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5fbb69812e49f9ecbf3e27483001956ba9f5892&vid=f5fbb69812e49f9ecbf3e27483001956ba9f5892&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR