Epicatechin(4b-8)epicatechin ≥90% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2R,2'R,3R,3'R,4R)-2,2'-Bis(3,4-dihydroxyphenyl)-[4,8'-bichromane]-3,3',5,5',7,7'-hexaol, Procyanidin B2

0209845MG
100263-474EA 405.38 USD
100263-474
Epicatechin(4b-8)epicatechin ≥90% (by HPLC), powder
Epicatechin(4b-8)epicatechin

  • Beige powder
  • Store at 2-8 °C, To be kept away from light

Formula: C₃₀H₂₆O₁₂
MW: 578.53 g/mol
MDL Number: MFCD01861513
CAS Number: 29106-49-8
UN: 12352100
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR