(-)-Epigallocatechin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2R,3R)-2-(3,4,5-Trihydroxyphenyl)chromane-3,5,7-triol, 3,5,7,3',4',5'-Hexahydroxylflavan

020979S10MG
100263-472EA 228.85 USD
100263-472
(-)-Epigallocatechin ≥98%, powder
(-)-Epigallocatechin

  • White powder
  • Store at 2-8 °C, Away from light

Formula: C₁₅H₁₄O₇
MW: 306.27 g/mol
Melting Pt: 219…221 °C
MDL Number: MFCD00075939
CAS Number: 970-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ba45e8409145310f66a1946a8b982784987f47d&vid=8ba45e8409145310f66a1946a8b982784987f47d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR