Loganin ≥90%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (1S,4aS,6S,7R,7aS)-Methyl 6-hydroxy-7-methyl-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylate

020203250MG
100263-384EA 741.46 USD
100263-384
Loganin ≥90%
Loganin
Formula: C₁₇H₂₆O₁₀
MW: 390.39 g/mol
MDL Number: MFCD00075645
CAS Number: 18524-94-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41a177d7cd73a54bf54798a3d7df1145e40adb56&vid=41a177d7cd73a54bf54798a3d7df1145e40adb56&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec4afb6692d9e3c09d9c2a4250a092a09e93f714&vid=ec4afb6692d9e3c09d9c2a4250a092a09e93f714&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR