Ginsenoside Rb1 ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2S,3R,4S,5S,6R)-2-(((2R,3R,4S,5S,6R)-4,5-Dihydroxy-2-(((3S,5R,8R,9R,10R,12R,13R,14R,17S)-12-hydroxy-4,4,8,10,14-pentamethyl-17-((S)-6-methyl-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)hept-5-en-2-yl)hexadecahydro-1H-cyclopenta[a]phenanthren-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

020105S5MG
100263-372EA 204 USD
100263-372
Ginsenoside Rb1 ≥98%, powder
Ginsenoside Rb1

  • White powder
  • To be kept in a desiccator

Formula: C₅₄H₉₂O₂₃
MW: 1109.3 g/mol
MDL Number: MFCD00133367
CAS Number: 41753-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01a53e6da619c3f67ce8c0557f7112641e37dd8f&vid=01a53e6da619c3f67ce8c0557f7112641e37dd8f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR