7-Methoxy-1,4-benzothiazin-3-one ≥98%, crystalline solid

Supplier: INDOFINE CHEMICAL COMPANY MS

250241GM
100263-300EA 49.69 USD
100263-300
7-Methoxy-1,4-benzothiazin-3-one ≥98%, crystalline solid
7-Methoxy-1,4-benzothiazin-3-one

Pale-pink crystalline solid


Store in a cool, dry place

Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Melting Pt: 180…182 °C
MDL Number: MFCD02660686
CAS Number: 22726-30-3
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1f165c5c049ac894a68d5506d1e90a3d455767a&vid=e1f165c5c049ac894a68d5506d1e90a3d455767a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR