8-Bromo-6-chloro-3(4'-methoxyphenyl)-4-methylcoumarin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19183250MG
100263-156EA 49.69 USD
100263-156
8-Bromo-6-chloro-3(4'-methoxyphenyl)-4-methylcoumarin ≥98%, powder
8-Bromo-6-chloro-3(4'-methoxyphenyl)-4-methylcoumarin

Cream colored shining powder


Store in a cool, dry place

Formula: C₁₇H₁₂BrClO₃
MW: 379.63 g/mol
Melting Pt: 184…185 °C
MDL Number: MFCD02660604
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92188eb2a77d76dc307e903c7b86c92e97f97295&vid=3816530c39e5ece327dca8a4cf30b61bb1a08420&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR