D-(+)-Galactose ≥98% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

024117100GM
100263-048EA 128.15 USD
100263-048
D-(+)-Galactose ≥98% (by HPLC), crystalline powder
D(+)-Galactose

  • White crystalline powder
  • Store in a cool, dry place

Formula: C₆H₁₂O₆
Melting Pt: 168…170 °C
MDL Number: MFCD00151230
CAS Number: 59-23-4
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c99dd90af95f0c58909fa54f371e00cf5d660ff&vid=0c99dd90af95f0c58909fa54f371e00cf5d660ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR