(-)-Epigallocatechin gallate ≥98% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG

020981S10MG
100262-980EA 117.69 USD
100262-980
(-)-Epigallocatechin gallate ≥98% (by HPLC), powder
(-)-Epigallocatechin gallate

  • Cream Powder
  • Store at 2-8 °C, Away from light

Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
Melting Pt: 220…229 °C
MDL Number: MFCD00075940
CAS Number: 989-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86419409c754e08905cf41c6f50547f1d9f8d9d6&vid=16ac89e5c7e03dcf75cca2843f96f496ba17656c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR