Pelargonidin-3,5-di-O-glucoside chloride ≥90% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

0209035MG
100262-974EA 387.08 USD
100262-974
Pelargonidin-3,5-di-O-glucoside chloride ≥90% (by HPLC)
Pelargonidin-3,5-di-O-glucoside chloride
Formula: C₂₇H₃₁O₁₅Cl
MDL Number: MFCD00017590
CAS Number: 17334-58-6
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8916bbdb9aed336080cf953f677ce795190c6b13&vid=8916bbdb9aed336080cf953f677ce795190c6b13&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87a74b9c9f9237368d1dc62ec511ce96f7e38037&vid=87a74b9c9f9237368d1dc62ec511ce96f7e38037&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR