Tissue-Tek® AutoTEC® a120 Automated Embedder, Sakura Finetek
Supplier: Sakura Finetek
7090
10025-432EA 296311.91 USD
10025-432
Tissue-Tek® AutoTEC® a120 Automated Embedder, Sakura Finetek
Tissue Embedders
Automated tissue embedder which utilizes the Paraform™ sectionable cassette system.
  • Barcode reader for track and trace of the Paraform cassette, allowing full connectivity with middleware and laboratory information systems
  • Heated input door and SMARTair™ technology removes excess paraffin from the cassettes to produce clean blocks that eliminates tedious scraping
  • 15" color touch screen with all operational information showing status of process, alerts with paraffin reservoir level, workload statistics
  • Specimen orientation is locked from gross dissection to microtomy
Continuous loading of specimens with four input doors of 20 cassettes each, achieves a rate of up to 120 cassettes per hour.
Certifications: UL 61010-1, IEC 61010-1, IEC 61010-2-010, IEC 61010-2-101.
Ordering information: Accessories include Six 20-cassette magazines; Four cassette magazine retainers; Two output doors; One paraffin inner tray #1, Two paraffin inner tray #2.