Inspection Mirror, Industrial Physics
Supplier: GREAT LAKES GAGES, LLC
LD3004
10025-270EA 57.62 USD
10025-270
Inspection Mirror, Industrial Physics
Mirrors
Inspection mirror spots that are difficult to inspect are even more difficult to blast or coat.
  • Ideal for visual inspection of hard-to-reach spots
  • With balljoint and pocket clip
With a simple inspection mirror a visual inspection behind stiffeners, under- or on top of I-beams or other places is easy to perform.