AimStep® Cotinine Test, Germaine® Laboratories
Supplier: Germaine Laboratories
40010
10024-940EA 2.31 USD
10024-940
AimStep® Cotinine Test, Germaine® Laboratories
Drug Tests
AimStep® Cotinine is a rapid qualitative test for the detection of cotinine in urine
Results are available in five minutes. Each test includes an internal procedural control.
The test is designed to detect cotinine at a concentration of 200ng/mL in urine. Each test is individually packed in a foil pouch.