AIM™ CRP Test, Germaine Laboratories
Supplier: Germaine Laboratories
32100
10024-410BX 46.74 USD
10024-410
AIM™ CRP Test, Germaine Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
This latex agglutination test is designed for the qualitative and semi-quantitative determination of C-Reactive Protein in non-diluted serum
This test is recommended for professional use.