Urine Analyzers, Germaine Laboratories
Supplier: Germaine Laboratories
51110
10024-378EA 5367.5 USD
10024-378
Urine Analyzers, Germaine Laboratories
Clinical Diagnostic Supplies Clinical Analysers
Automated analyzer designed for the semi-quantitative detection of up to 10 analytes in urine
The Urine Analyzer can be set to continous or routine mode. Languages include English, Spanish, and Portuguese. The unit can store up to 2,000 results, and is compatible with a wide range of LSI or EMR systems. Automatic calibration ensures accurate results.
Analyzers will read AimStrip® 10-SG and AimStrip® US-4OB. The high output analyzer can support up to 500 tests per hour, and features a large touchscreen for easy naviagation.
Certifications: CLIA waived.
Ordering information: Analyzers are supplied with power cord, user's manual, and two printer paper rolls. A barcode reader is also available separately.