AimStrip® Adulteration 7 Test, Germaine Laboratories
Supplier: Germaine Laboratories
40007
10024-370PK 38.8 USD
10024-370
AimStrip® Adulteration 7 Test, Germaine Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
These urine Dipsticks are designed as a semi-quantitative, color comparison screen for the detection of Creatinine, Nitrite, Glutaraldehyde, pH, Specific Gravity, and Oxidants-Pyridinium Chlorochromate (PCC) in human urine
Results are available within 2 to 5 minutes. Results are determined by comparing the reagent pads on the dipstick to a color chart provided on the bottle. Test includes 25 dipsticks per bottle.
This test provides a preliminary screen only, and is intended for forensic use only.