Nalgene® Right-To-Understand Safety Wash Bottles, Thermo Fisher Scientific
Supplier: Thermo Fisher Scientific
2429-0504
10024-300CS 1008.37 USD
10024-300 10024-302 10024-304 10024-306 10024-308 10024-804
Nalgene® Right-To-Understand Safety Wash Bottles, Thermo Fisher Scientific
Bottles Wash Bottles
FEP bottle with ETFE dispensing cap.
  • Greater awareness of hazards, resulting in safer use of chemicals in the workplace; detailed safety information
  • at your fingertips
  • 100% GHS information (no mixing of systems): wash bottles are labelled with the same warning system as is
  • used on purchased solvents
  • Chemically inert (low leachable) making it suitable for storing high purity solvent
  • Autoclavable
Bottles are pre-labelled with chemical identification and safe handling information following the GHS specifications as applicable to secondary laboratory containers. Wash bottles are ready to use when you transfer harsh laboratory chemicals from manufacturer's containers.
Permanently laser etched with the solvent safety info, so the information can never come off or become illegible throughout the life of the bottle (FEP wash bottles are known to be difficult to label due to their slippery surface. Markers, wax pencils, and labels won't stick).