Manual stereotaxic injector, STOELTING MS
Supplier: Stoelting
51639
100231-302EA 1431.92 USD
100231-302
Manual stereotaxic injector, STOELTING MS
Syringes Syringes for Chromatography
Manual Stereotaxic Injector provides an economical means of injecting/withdrawing a calibrated volume with simple, precise manual control
Mounted directly on a stereotaxic frame, it allows precise injection directly through the needle into the tissue. Injection directly through a needle on the syringe avoids the compliance issues that arise when injections are forced through plastic tubing. Volumes as small as 200 picoliters may be infused or withdrawn.