Cordless Micro Drill, Stoelting
Supplier: Stoelting
58610
100230-730EA 719.23 USD
100230-730
Cordless Micro Drill, Stoelting
Tool Sets
The Cordless Micro Drill is designed for research applications that require surgical bits
  • Speed is 14400 RPM
  • Can function for eight hours on a single charge
  • Powered by a rechargeable 6-volt nickel cadmium battery
Made of lightweight aluminum alloy for balance and control. The Micro Drill can function for up to eight hours on a single charge and is powered by a rechargeable, 6-volt cadmium battery,
Delivery information: The kit includes a hard plastic case with die-cut foam for convenient storage, a recharger, and five carbide steel burrs with ball sizes of 0.6 mm, 0.8 mm, 1.0 mm, 1.2 mm and 1.6 mm. Carbide burr drill bits have a shaft diameter of 2.3 mm and are 4.4 cm in length. No special tools are necessary to change burrs.