Buchi® F-308 Recirculating Chillers
Supplier: Buchi
11F30802
10023-988EA 11533.05 USD
10023-988
Buchi® F-308 Recirculating Chillers
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The F-100 recirculating chillier is a compact product designed to work with laboratory equipment such as rotary evaporators, parallel evaporators, kjeldahl, and extraction products.
  • ECO-mode: saves energy and reduces heat emission by switching into a stand-by mode when not in use
  • No water consumption
  • Maximizes distillation capacity while reducing solvent emissions