Rotavapor® R-300M Rotary Evaporators Standalone, BUCHI
Supplier: Buchi
11R300152C102 11R300152V002 11R300152V102 11R300152C002
10023-914EA 7944.36 USD
10023-914 10024-012 10024-014 10024-016
Rotavapor® R-300M Rotary Evaporators Standalone, BUCHI
Evaporators Rotary Evaporators
The R-300 meets the highest expectations in convenience and versatility.
  • Intuitive user interaction
  • Precise operation control
  • 5 L flask capacity
  • Ergonomic handle
  • Combi-clip
  • Additional glass assemblies and configurations
The R-300 Standalone exhibits a central integrated display of heating bath temperature, rotation speed and lift position on the heating bath base. The rotation speed and heating bath temperature are precisely controlled via two intuitively positioned control knobs. The patented multipurpose Combi-clip with snap lock-mechanism provides for quick flask connection and detachment with one hand.
Its modular design allows extension of the Rotavapor® R-300 to a fully integrated system where a central interface regulates each component.
Certifications: CE and CSA compliant.
Ordering information: All accessories and connections are included for out of the box usage. Optional 1L bath, safety shield, >1L evaporation flasks, different joint size glassware, valves for in-house cooling and vacuum are available as accessories (sold separately). Additional glass assemblies and configurations are available. Please contact customer service for more information.