180 QT Portable Analyzer, Pointe Scientific

Supplier: HORIBA INSTRUMENTS INCORPORATED
90-P4504-QT
10023-514EA 12294.61 USD
10023-514
180 QT Portable Analyzer, Pointe Scientific
Clinical Diagnostic Supplies Clinical Analysers
A compact, microprocessor controlled analyzer that can read and calculate results of endpoint, kinetic, initial rate and colorimetric assays. Ideal for teaching future laboratory professionals the fundamentals of clinical chemistry testing

  • Portable
  • Lightweight
  • Light Source: Tungsten lamp with lamp-saver feature
  • Standard Wavelengths: 340, 405, 505, 545, 580 and 630 nm
  • Display: Interactive touch screen 3.5” LCD color graphic display
  • Printer: Thermal dot matrix with graphic capability

Microprocessor used are eZ80, 64K EEPROM. It Stores all parameters including wavelengths, calculations, unit codes, linear and normal ranges, rate timing, standard values, test names, and previous standard curve.

Ordering information: For research and education use only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR