Magnesium Reagent Set, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
OM929-300
10022-808EA 140.67 USD
10022-808 10022-810 10022-812 10023-496
Magnesium Reagent Set, Pointe Scientific
Assays Protein Assays
The Magnesium Reagent Set is for the quantitative determination of magnesium in serum.
  • Quantitative determination of magnesium in serum
  • For in vitro diagnostic use only
  • Serum magnesium ions react with Xylidyl Blue in an alkaline solution to produce a red complex that is measured spectrophotometrically
The Magnesium Reagent Set is for the quantitative determination of magnesium in serum
The Hitachi 300 mL set is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are six 25 mL R1 Reagent bottles and six 25 mL R2 Reagent bottles in this kit.
The Hitachi 400 mL set is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are four 50 mL R1 Reagent bottles and four 50 mL R2 Reagent bottles in this kit.
The Olympus reagent set is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are six 25 mL R1 Reagent bottles and six 25 mL R2 Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
A NIST-traceable magnesium standard or serum based calibrator and control sera with known normal and abnormal magnesium values.