Cholesterol Liquid Reagent Set, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
OC910-780
10022-720EA 96.97 USD
10022-720 10022-722 10022-724 10023-478
Cholesterol Liquid Reagent Set, Pointe Scientific
Assays Immunological Assays
The Cholesterol Liquid Reagent Set is used for the quantitative determination of Total Cholesterol in serum.
  • Quantitative determination of total cholesterol in serum
  • Free cholesterol is oxidized by cholesterol oxidase to release hydrogen peroxide
  • For in vitro diagnostic use only
The Cholesterol Liquid Reagent Set is used for the quantitative determination of Total Cholesterol in serum
The Olympus reagent kit is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are (12) 65 mL bottles in this kit.
Certifications: CE Certified, Health Canada
Ordering information: Materials Required but not Provided:
Aqueous standards can be used to calibrate the procedure or an appropriate serum calibrator as well as Serum controls with known normal and elevated values.