autoHDL Cholesterol Reagent, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
OH945-480
10022-764EA 154.5 USD
10022-764 10022-766 10022-768 10022-770 10023-466
autoHDL Cholesterol Reagent, Pointe Scientific
Clinical Diagnostic Supplies Clinical Reagents and Kits
The autoHDL Cholesterol Reagent is for the quantitative determination of high-density lipoprotein cholesterol in human serum or plasma.
  • Quantitative determination of high-density lipoprotein cholesterol in human serum or plasma
  • Homogeneous method for directly measuring serum HDL-C levels
  • No need for any off-line pretreatment or centrifugation steps
The autoHDL Cholesterol Reagent is for the quantitative determination of high-density lipoprotein cholesterol in human serum or plasma
The Hitachi 480 mL Reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are six 60 mL R1 Reagent bottles and six 20 mL R2 Reagent bottles in this kit.
The Hitachi 240 mL Reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are six 30 mL R1 Reagent bottles and three 30 mL R2 Reagent bottles in this kit.
The Olympus Reagent is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are six 60 mL R1 Reagent bottles and six 20 mL R2 Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
An autoHDL/LDL Cholesterol Calibrator and HDL cholesterol controls.