Albumin Reagent Set, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
OA902-792
10022-504EA 94.54 USD
10022-504 10022-506 10022-632 10022-634 10023-456
Albumin Reagent Set, Pointe Scientific
Assays Protein Assays
The Albumin Reagent Set is used for the quantitative determination of Albumin in serum.
  • Quantitative determination of Albumin in serum
  • Increase in blue-green color
  • Standard is used to calibrate the determination of unknown specimens
Used for the quantitative determination of Albumin in serum
The Olympus Albumin Reagent is used on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. This reagent is measured at 600/700nm.
Albumin is bound by the BCG dye to produce an increase in the blue-green color measured at 630 nm. The color increase is proportional to the concentration of albumin present.
Certifications: CE Compliant
Ordering information: Materials Required but not Provided: Either Chemistry Calibrator or Albumin Standard; as well as both Normal and Abnormal Control Sera with known Albumin Concentrations Required.