Glycerine solution 50%, sterile

Supplier: TEKNOVA INC

Synonyms: Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol

G1799 G1798
100217-154EA 99.43 USD
100217-154 100217-622 100220-042
Glycerine solution 50%, sterile
Glycerine

Glycerol has many uses including increasing viscosity and density of solutions.

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Flash Pt: 177
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR