LB Lennox Broth Bottled Media, Sterile, Teknova
Supplier: TEKNOVA INC
L5000
100220-018EA 131.05 USD
100220-018
LB Lennox Broth Bottled Media, Sterile, Teknova
Media for Microbiology Prepared Media for Microbiology
LB Lennox version of LB Broth
  • Contains less salt than the most common formulation of LB, known as LB Miller
  • Sterile
LB Lennox is commonly used for cloning in recombinant E. coli and selection with salt-sensitive antibiotics such as Blasticidin, CloNat, Hygromycin, Puromycin and Zeocin. Ingredients: 1.0% Tryptone (10.0g/L), 0.5% Yeast Extract (5.0g/L), 0.5% NaCl (5.0g/L). Ambient shipping conditions. Store at room temperature or 2 to 8°C.
For the growth of E. coli and other enteric bacteria.