Two-Piece Biopsy Cassettes, Azer Scientific
Supplier: VWR International
ES3014-W
10022-302CS 139.67 USD
10022-302 10022-348
Two-Piece Biopsy Cassettes, Azer Scientific
Embedding Cassettes Biopsy Processing Cassettes
These cassettes are molded from a special, high-density polymer that is resistant to the chemical actions of solvents.
  • Three frosted writing areas
  • High-density polymer resistant to chemical actions
  • Relocking lid with no risk of specimen loss
  • Two-piece design
Lids can be snapped on and off as often as necessary and relock securely.
The cassettes include three frosted labeling areas.
Packaging: 500 per case