Foam Biopsy Pads, Globe Scientific
Supplier: Globe Scientific
190195
10022-166PK 58.47 USD
10022-166
Foam Biopsy Pads, Globe Scientific
Biopsy Pads
The Globe Scientific foam biopsy pads are used to hold biopsies in place and prevent their loss during processing
They are produced from a special polyester urethane foam, which is verified for consistency throughout in order to ensure optimum solvent flow. Simply sandwich the biopsy sample between two pads and insert into a cassette for processing.