Viscosity Standards, Silicone Oils, Cannon Instruments
Supplier: CANNON INSTRUMENT CO
9727-Z10.016 9727-Z10.032 9727-Z10.128 9727-Z10.5 9727-Z14.016 9727-Z14.032 9727-Z14.128 9727-Z14.5 9727-Z18.016 9727-Z18.032 9727-Z18.128 9727-Z18.5 9727-Z22.016 9727-Z22.032 9727-Z22.128 9727-Z22.5 9727-Z26.016 9727-Z26.032 9727-Z26.128 9727-Z26.5 9727-Z30.016 9727-Z30.032 9727-Z30.128 9727-Z30.5 9727-Z34.016 9727-Z34.032 9727-Z34.128 9727-Z42.032 9727-Z42.128 9727-Z42.5 9727-Z46.016 9727-Z46.032 9727-Z46.128 9727-Z46.5 9727-Z50.016 9727-Z50.032 9727-Z50.128 9727-Z50.5 9727-Z34.5 9727-Z38.016 9727-Z38.032 9727-Z38.128 9727-Z38.5 9727-Z42.016
10021-850EA 238 USD
10021-850 10021-852 10021-854 10021-856 10021-858 10021-860 10021-862 10021-864 10021-866 10021-868 10021-870 10021-872 10021-874 10021-876 10021-878 10021-880 10021-882 10021-884 10021-886 10021-888 10021-890 10021-892 10021-894 10021-896 10021-898 10021-900 10021-902 10021-904 10021-906 10021-908 10021-910 10021-912 10021-914 10021-916 10021-918 10021-920 10021-922 10021-924 10022-098 10022-100 10022-102 10022-104 10022-106 10022-108
Viscosity Standards, Silicone Oils, Cannon Instruments
Standards Viscometry Standards
CANNON silicone standards are intended for calibration of brookfield type rotational viscometers.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Certifications: Traceable to NIST.