Terrific Broth Complete, Bottled Media, Teknova
Supplier: TEKNOVA INC
T7060
100219-866EA 134.69 USD
100219-866
Terrific Broth Complete, Bottled Media, Teknova
Media for Microbiology Prepared Media for Microbiology
Used to obtain high yields of plasmid DNA from E.coli.
  • 500 mL and 1 Liter
  • Sterile
Contains 1.2% Tryptone, 2.4% yeast extract, 0.4% glycerol, 72 mM K2HPO4, and 17 mM KH2PO4. Ambient shipping conditions. Store at room temperature.