Carbenicillin

Supplier: TEKNOVA INC

L8185
100219-764EA 133.43 USD
100219-764
Carbenicillin
Carbenicillin

1.0% Tryptone, 0.5% yeast extract, 1.0% NaCl with 100 g/mL carbenicillin.Applications: Suitable for use with carbenicillin (ampicillin) resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC, etc.

Formula: C₁₇H₁₈N₂O₆S
CAS Number: 4697-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR