LB Broth with 50 µg/mL Kanamycin Bottled Media, Sterile, Teknova
Supplier: TEKNOVA INC
L8145
100219-756EA 136.35 USD
100219-756
LB Broth with 50 µg/mL Kanamycin Bottled Media, Sterile, Teknova
Media for Microbiology Prepared Media for Microbiology
For the maintenance and propagation of kanamycin resistant strains.
  • 1 Liter
  • Sterile
Contains 1.0% Tryptone, 0.5% yeast extract, 1.0% NaCl, 50µg/ml kanamycin. Ship on ice. Store frozen.