Water, Molecular Biology Grade, PCR certified DNase/RNase tested, sterile

Supplier: TEKNOVA INC

Synonyms: Dihydrogen oxide

W3350 W3345 W3330 W3335 W3340
100216-650EA 84.84 USD
100216-650 100220-288 100219-594 76423-626 76423-628
Water, Molecular Biology Grade, PCR certified DNase/RNase tested, sterile
Water

Molecular Biology Grade Water is PCR certified negative for DNAse and RNAse (sensitivity to femtomoles) and is negative for non-specific Endonuclease, Exonuclease and Protease activity.


  • Sterile

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

DNase tested
RNase tested
Protease tested
PCR tested


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR