N-Lauroylsarcosine sodium salt 10%
Supplier: TEKNOVA INC
|
Warning
|
Synonyms:
Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate
S3376
100218-364EA
197.49
USD
100218-364
N-Lauroylsarcosine sodium salt 10%
N-Lauroylsarcosine sodium salt
Sarkosyl (sodium lauroyl sarcosinate) is an anionic detergent that is less denaturing than SDS. Uses include solubilization of membranes and cell lysis for extraction of RNA.
Formula:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa MW: 293.38 g/mol |
MDL Number:
MFCD00042728 CAS Number: 137-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...