MOPS/SDS, 20X, Running Buffer, Teknova
Supplier: TEKNOVA INC
M1089
550000-032EA 568.65 USD
550000-032 100217-904
MOPS/SDS, 20X, Running Buffer, Teknova
Buffers Electrophoresis Buffers Running Buffers
MOPS/SDS running buffer is designed for protein electrophoresis.
  • Sterile
20X liquid concentrate contains 1 M MOPS, 1 M Tris, 20.5 mM EDTA and 69.3 mM SDS.