Teknova SOB Broth
Supplier: TEKNOVA INC
S0210
100216-662EA 119.61 USD
100216-662
Teknova SOB Broth
Media for Microbiology Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.