Tween® 20 (Polysorbate) 10% in aqueous solution

Supplier: TEKNOVA INC

T0710
100216-360EA 127.62 USD
100216-360
Tween® 20 (Polysorbate) 10% in aqueous solution
Tween® 20 (Polysorbate)

Tween®20 is a polyoxyethylene sorbitol ester that belongs to the polysorbate family. It is a nonionic detergent having a molecular weight of 1,225 daltons, assuming 20 ethylene oxide units, 1 sorbitol, and 1 lauric acid as the primary fatty acid.


Caution: TWEEN is a registered trademark of Croda International PLC.

Formula: C₅₈H₁₁₄O₂₆
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR