Zinc sulfide, crystal disk for spectrophotometry, polished
Supplier: INTERNATIONAL CRYSTAL LABS MS
Synonyms:
Zinc (II) sulphide, Zinc (II) sulfide
0002I-1858
0002I-4591
0002I-151
0002I-152
0002I-1011
0002I-77
0002I-464
100211-352EA
428.23
USD
100211-352
100214-462
100212-948
100215-600
100211-670
100216-086
100216-122
Zinc sulfide, crystal disk for spectrophotometry, polished
Zinc sulphide
Zinc Sulfide (ZnS) is used as a transmission window for IR spectroscopy. Zinc Sulfide (ZnS) is or has been produced under the trademarks Cleartran® and Irtran® .
Formula:
SZn MW: 97.46 g/mol Melting Pt: 1064 Density: 4.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011301 CAS Number: 1314-98-3 EINECS: 215-251-3 Merck Index: 12,10294 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...