Zinc sulfide, crystal random cuttings for spectrophotometry

Supplier: INTERNATIONAL CRYSTAL LABS MS

Synonyms: Zinc (II) sulphide, Zinc (II) sulfide

0002-4534
100215-356EA 1344.62 USD
100215-356
Zinc sulfide, crystal random cuttings for spectrophotometry
Zinc sulphide

Zinc Sulfide (ZnS) is used as a transmission window for IR spectroscopy. Zinc Sulfide (ZnS) is or has been produced under the trademarks Cleartran® and Irtran®.

Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064 °C
Density: 4.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 12,10294

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR