Calcium fluoride, crystal disk for spectrophotometry, polished

Supplier: INTERNATIONAL CRYSTAL LABS MS
Warning

0002D-410 0002D-181 0005D-4488 0002D-548 0002D-403 0002D-1455 0002D-1457 0002D-1006 0002D-259
100212-968EA 171 USD
100212-968 100211-850 100214-460 100215-426 100214-062 100211-884 100211-714 100213-476 100214-562
Calcium fluoride, crystal disk for spectrophotometry, polished
Calcium fluoride

International Crystal Labs celebrated its 40th anniversary in 2002. They have produced quality crystal optics for spectroscopy and CO2 lasers since 1962. ICL operates a vertically integrated crystal optics production plant and grow their own crystals.

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500
Melting Pt: 1418
Density: 3.18 g/cm³ (20 °C)
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR