Calcium fluoride, crystal rectangle for spectrophotometry, polished, drilled

Supplier: INTERNATIONAL CRYSTAL LABS MS
Warning

0002D-92
100214-554EA 247 USD
100214-554
Calcium fluoride, crystal rectangle for spectrophotometry, polished, drilled
Calcium fluoride

International Crystal Labs celebrated its 40th anniversary in 2002. They have produced quality crystal optics for spectroscopy and CO2 lasers since 1962. ICL operates a vertically integrated crystal optics production plant and grow their own crystals.

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500
Melting Pt: 1418
Density: 3.18 g/cm³ (20 °C)
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR